Assay Details
| Gene Symbol: LOC101712192 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004855836.3 | XP_004855893.1 | 4 - 5 | 744 | 112 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101712192| Gene Symbol: LOC101712192 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004855836.3 | XP_004855893.1 | 4 - 5 | 744 | 112 | |
Set Membership: |
|