Assay Details
| Gene Symbol: March2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004865410.3 | XP_004865467.2 | 3 - 4 | 444 | 86 | |
| XM_004865412.2 | XP_004865469.1 | 3 - 4 | 528 | 86 | ||
| XM_004865413.3 | XP_004865470.1 | 3 - 4 | 1015 | 86 | ||
More Information
Set Membership: |
|

March2
Genomic Map