Assay Details
| Gene Symbol: Sigmar1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004838438.3 | XP_004838495.1 | 1 - 2 | 246 | 77 | |
| XM_004838439.3 | XP_004838496.2 | 1 - 2 | 256 | 77 | ||
More Information
Set Membership: |
|

Sigmar1
Genomic Map