Assay Details
| Gene Symbol: Clic5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004846430.3 | XP_004846487.1 | 3 - 4 | 839 | 61 | |
| XM_004846431.3 | XP_004846488.1 | 3 - 4 | 575 | 61 | ||
More Information
Set Membership: |
|

Clic5
Genomic Map