Assay Details
| Gene Symbol: Ptges | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004849133.3 | XP_004849190.1 | 1 - 2 | 197 | 70 | |
| XM_021261889.1 | XP_021117548.1 | 1 - 2 | 207 | 70 | ||
More Information
Set Membership: |
|

Ptges
Genomic Map