Assay Details
| Gene Symbol: Pofut1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004840581.3 | XP_004840638.1 | 4 - 5 | 627 | 65 | |
| XM_021254891.1 | XP_021110550.1 | 4 - 5 | 863 | 65 | ||
| XM_021254892.1 | XP_021110551.1 | 4 - 5 | 937 | 65 | ||
More Information
Set Membership: |
|

Pofut1
Genomic Map