Assay Details
| Gene Symbol: Rnase7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004865031.3 | XP_004865088.1 | 2 | 1056 | 84 | |
| XM_021244575.1 | XP_021100234.1 | 3 | 1110 | 84 | ||
| XM_021244576.1 | XP_021100235.1 | 3 | 1132 | 84 | ||
| XM_021244577.1 | XP_021100236.1 | 3 | 1092 | 84 | ||
| XM_021244578.1 | XP_021100237.1 | 3 | 1114 | 84 | ||
| XM_021244579.1 | XP_021100238.1 | 3 | 1090 | 84 | ||
| XM_021244580.1 | XP_021100239.1 | 3 | 1068 | 84 | ||
| XM_021244581.1 | XP_021100240.1 | 2 | 1034 | 84 | ||
More Information
Set Membership: |
|

Rnase7
Genomic Map