Assay Details
| Gene Symbol: LOC101696396 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013075040.2 | XP_012930494.2 | 7 - 8 | 769 | 82 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101696396| Gene Symbol: LOC101696396 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013075040.2 | XP_012930494.2 | 7 - 8 | 769 | 82 | |
Set Membership: |
|