Assay Details
| Gene Symbol: LOC101726493 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004865937.2 | XP_004865994.1 | 2 - 3 | 319 | 59 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101726493| Gene Symbol: LOC101726493 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004865937.2 | XP_004865994.1 | 2 - 3 | 319 | 59 | |
Set Membership: |
|