Assay Details
| Gene Symbol: Lsm6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004868920.3 | XP_004868977.1 | 2 - 3 | 224 | 92 | |
| XM_004868921.2 | XP_004868978.1 | 2 - 3 | 223 | 92 | ||
| XM_013067120.2 | XP_012922574.1 | 2 - 3 | 224 | 92 | ||
| XM_021247680.1 | XP_021103339.1 | 2 - 3 | 224 | 92 | ||
| XM_021247681.1 | XP_021103340.1 | 2 - 3 | 224 | 92 | ||
| XM_021247682.1 | XP_021103341.1 | 2 - 3 | 224 | 92 | ||
| XM_021247683.1 | XP_021103342.1 | 2 - 3 | 224 | 92 | ||
More Information
Set Membership: |
|

Lsm6
Genomic Map