Assay Details
| Gene Symbol: Bdkrb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004837013.2 | XP_004837070.1 | 1 - 2 | 225 | 80 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_004837013.2 transcripts.
Set Membership: |
|

Bdkrb1
Genomic Map