Assay Details
| Gene Symbol: Slc25a24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004853592.2 | XP_004853649.1 | 7 - 8 | 923 | 64 | |
| XM_021265582.1 | XP_021121241.1 | 7 - 8 | 901 | 64 | ||
More Information
Set Membership: |
|

Slc25a24
Genomic Map