Assay Details
| Gene Symbol: Rps19bp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004845566.3 | XP_004845623.1 | 2 - 3 | 230 | 67 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rps19bp1| Gene Symbol: Rps19bp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004845566.3 | XP_004845623.1 | 2 - 3 | 230 | 67 | |
Set Membership: |
|