Assay Details
| Gene Symbol: Syngr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004860817.2 | XP_004860874.1 | 1 - 2 | 156 | 83 | |
| XM_021241043.1 | XP_021096702.1 | 1 - 2 | 156 | 83 | ||
More Information
Set Membership: |
|

Syngr2
Genomic Map