Assay Details
| Gene Symbol: Elof1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004865607.3 | XP_004865664.1 | 3 - 4 | 336 | 58 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Elof1| Gene Symbol: Elof1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004865607.3 | XP_004865664.1 | 3 - 4 | 336 | 58 | |
Set Membership: |
|