Assay Details
| Gene Symbol: Ndufc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004869676.3 | XP_004869733.1 | 2 - 3 | 473 | 116 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ndufc2| Gene Symbol: Ndufc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004869676.3 | XP_004869733.1 | 2 - 3 | 473 | 116 | |
Set Membership: |
|