Assay Details
| Gene Symbol: Cct6a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004839969.2 | XP_004840026.1 | 4 - 5 | 617 | 93 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cct6a| Gene Symbol: Cct6a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004839969.2 | XP_004840026.1 | 4 - 5 | 617 | 93 | |
Set Membership: |
|