Assay Details
| Gene Symbol: LOC101716142 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004848216.3 | XP_004848273.1 | 4 - 5 | 905 | 87 | |
| XM_004848218.2 | XP_004848275.1 | 3 - 4 | 797 | 87 | ||
| XM_021261199.1 | XP_021116858.1 | 4 - 5 | 1072 | 87 | ||
More Information
Set Membership: |
|

LOC101716142
Genomic Map