Assay Details
| Gene Symbol: Rps14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004854423.3 | XP_004854480.1 | 1 - 2 | 201 | 79 | |
| XM_021266221.1 | XP_021121880.1 | 1 - 2 | 202 | 79 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_004854423.3, XM_021266221.1 transcripts.
Set Membership: |
|

Rps14
Genomic Map