Assay Details
| Gene Symbol: Cst6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004852343.3 | XP_004852400.1 | 1 - 2 | 320 | 96 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cst6| Gene Symbol: Cst6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004852343.3 | XP_004852400.1 | 1 - 2 | 320 | 96 | |
Set Membership: |
|