Assay Details
| Gene Symbol: Brk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004835327.2 | XP_004835384.1 | 1 - 2 | 196 | 74 | |
| XM_021260538.1 | XP_021116197.1 | 1 - 2 | 196 | 74 | ||
More Information
Set Membership: |
|

Brk1
Genomic Map