Assay Details
| Gene Symbol: Rtraf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004835538.2 | XP_004835595.1 | 6 - 7 | 667 | 90 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rtraf| Gene Symbol: Rtraf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004835538.2 | XP_004835595.1 | 6 - 7 | 667 | 90 | |
Set Membership: |
|