Assay Details
| Gene Symbol: Sdha | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004845101.3 | XP_004845158.1 | 12 - 13 | 1760 | 60 | |
| XM_004845102.3 | XP_004845159.1 | 12 - 13 | 1760 | 60 | ||
| XM_013072383.2 | XP_012927837.1 | 12 - 13 | 1757 | 60 | ||
More Information
Set Membership: |
|

Sdha
Genomic Map