Assay Details
| Gene Symbol: Kpna2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004870244.3 | XP_004870301.1 | 9 - 10 | 1484 | 65 | |
| XM_013067642.2 | XP_012923096.1 | 9 - 10 | 1547 | 65 | ||
More Information
Set Membership: |
|

Kpna2
Genomic Map