Assay Details
| Gene Symbol: Vps26c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004842385.2 | XP_004842442.1 | 4 - 5 | 520 | 85 | |
| XM_021256263.1 | XP_021111922.1 | 4 - 5 | 557 | 85 | ||
More Information
Set Membership: |
|

Vps26c
Genomic Map