Assay Details
| Gene Symbol: Rack1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004875125.3 | XP_004875182.1 | 3 - 4 | 536 | 105 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rack1| Gene Symbol: Rack1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004875125.3 | XP_004875182.1 | 3 - 4 | 536 | 105 | |
Set Membership: |
|