Assay Details
| Gene Symbol: LOC101708617 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004843418.3 | XP_004843475.1 | 1 - 2 | 214 | 72 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101708617| Gene Symbol: LOC101708617 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004843418.3 | XP_004843475.1 | 1 - 2 | 214 | 72 | |
Set Membership: |
|