Assay Details
| Gene Symbol: Rbmx2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004860063.3 | XP_004860120.1 | 4 - 5 | 408 | 87 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rbmx2| Gene Symbol: Rbmx2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004860063.3 | XP_004860120.1 | 4 - 5 | 408 | 87 | |
Set Membership: |
|