Assay Details
| Gene Symbol: Mdk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004851922.3 | XP_004851979.2 | 3 - 4 | 415 | 59 | |
| XM_004851923.2 | XP_004851980.1 | 3 - 4 | 355 | 59 | ||
More Information
Set Membership: |
|

Mdk
Genomic Map