Assay Details
| Gene Symbol: C1s | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004869483.3 | XP_004869540.1 | 10 - 11 | 1531 | 64 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

C1s| Gene Symbol: C1s | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004869483.3 | XP_004869540.1 | 10 - 11 | 1531 | 64 | |
Set Membership: |
|