Assay Details
| Gene Symbol: Rac1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004840439.3 | XP_004840496.1 | 4 - 5 | 398 | 79 | |
| XM_021254729.1 | XP_021110388.1 | 5 - 6 | 478 | 79 | ||
| XM_021254730.1 | XP_021110389.1 | 4 - 5 | 426 | 79 | ||
More Information
Set Membership: |
|

Rac1
Genomic Map