Assay Details
| Gene Symbol: Faap20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004863801.3 | XP_004863858.2 | 2 - 3 | 289 | 77 | |
| XM_004863802.3 | XP_004863859.1 | 2 - 3 | 324 | 77 | ||
| XM_004863804.3 | XP_004863861.1 | 2 - 3 | 208 | 77 | ||
| XM_013065102.2 | XP_012920556.2 | 2 - 3 | 289 | 77 | ||
| XM_021243388.1 | XP_021099047.1 | 2 - 3 | 664 | 77 | ||
More Information
Set Membership: |
|

Faap20
Genomic Map