Assay Details
| Gene Symbol: Gabra3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004871055.2 | XP_004871112.1 | 9 - 10 | 1210 | 74 | |
| XM_013067906.2 | XP_012923360.1 | 9 - 10 | 1357 | 74 | ||
More Information
Set Membership: |
|

Gabra3
Genomic Map