Assay Details
Gene Symbol: Arpp19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013074301.2 | XP_012929755.1 | 2 - 3 | 306 | 80 | |
XM_013074303.2 | XP_012929757.1 | 2 - 3 | 319 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
