Assay Details
| Gene Symbol: Pros1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021238816.1 | XP_021094475.1 | 11 - 12 | 1475 | 71 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pros1| Gene Symbol: Pros1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021238816.1 | XP_021094475.1 | 11 - 12 | 1475 | 71 | |
Set Membership: |
|