Assay Details
| Gene Symbol: LOC101720875 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004865918.3 | XP_004865975.1 | 3 - 4 | 276 | 73 | |
| XM_013065998.2 | XP_012921452.1 | 3 - 4 | 259 | 73 | ||
| XM_013065999.2 | XP_012921453.1 | 2 - 3 | 189 | 73 | ||
| XM_021244957.1 | XP_021100616.1 | 5 - 6 | 923 | 73 | ||
| XM_021244959.1 | XP_021100618.1 | 5 - 6 | 925 | 73 | ||
| XM_021244964.1 | XP_021100623.1 | 4 - 5 | 422 | 73 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_013065999.2 transcripts.
Set Membership: |
|

LOC101720875
Genomic Map