Assay Details
| Gene Symbol: LOC101709232 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004862205.3 | XP_004862262.1 | 1 - 2 | 110 | 81 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_004862205.3 transcripts.
Set Membership: |
|

LOC101709232
Genomic Map