Assay Details
| Gene Symbol: Pde6g | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004860936.3 | XP_004860993.1 | 2 - 3 | 1251 | 74 | |
| XM_021241254.1 | XP_021096913.1 | 2 - 3 | 1418 | 74 | ||
More Information
Set Membership: |
|

Pde6g
Genomic Map