Assay Details
| Gene Symbol: LOC110343857 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021259428.1 | XP_021115087.1 | 2 | 874 | 66 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC110343857| Gene Symbol: LOC110343857 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021259428.1 | XP_021115087.1 | 2 | 874 | 66 | |
Set Membership: |
|