Assay Details
| Gene Symbol: LOC101708221 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021250485.1 | XP_021106144.1 | 1 - 2 | 93 | 97 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_021250485.1 transcripts.
Set Membership: |
|

LOC101708221
Genomic Map