Assay Details
| Gene Symbol: LOC101708221 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013068425.2 | XP_012923879.2 | 8 - 9 | 1024 | 88 | |
| XM_021250485.1 | XP_021106144.1 | 8 - 9 | 837 | 88 | ||
More Information
Set Membership: |
|

LOC101708221
Genomic Map