Assay Details
| Gene Symbol: Mogat2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004863215.2 | XP_004863272.1 | 3 - 4 | 525 | 64 | |
| XM_021243298.1 | XP_021098957.1 | 3 - 4 | 525 | 64 | ||
| XM_021243299.1 | XP_021098958.1 | 3 - 4 | 525 | 64 | ||
More Information
Set Membership: |
|

Mogat2
Genomic Map