Assay Details
| Gene Symbol: Rnasel | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004862830.2 | XP_004862887.1 | 4 - 5 | 2024 | 76 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rnasel| Gene Symbol: Rnasel | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004862830.2 | XP_004862887.1 | 4 - 5 | 2024 | 76 | |
Set Membership: |
|