Assay Details
| Gene Symbol: LOC101721836 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021261540.1 | XP_021117199.1 | 2 - 3 | 323 | 80 | |
| XM_021261541.1 | XP_021117200.1 | 2 - 3 | 325 | 80 | ||
More Information
Set Membership: |
|

LOC101721836
Genomic Map