Assay Details
| Gene Symbol: LOC101721836 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021261540.1 | XP_021117199.1 | 3 - 4 | 391 | 90 | |
| XM_021261541.1 | XP_021117200.1 | 3 - 4 | 393 | 90 | ||
More Information
Set Membership: |
|

LOC101721836
Genomic Map