Assay Details
| Gene Symbol: Nrbf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004858257.3 | XP_004858314.1 | 3 - 4 | 358 | 58 | |
| XM_013077197.2 | XP_012932651.1 | 2 - 3 | 274 | 58 | ||
More Information
Set Membership: |
|

Nrbf2
Genomic Map