Assay Details
| Gene Symbol: Hamp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004858837.3 | XP_004858894.1 | 1 - 2 | 208 | 76 | |
| GenBank mRNA | KM486717.1 | - | 1 - 2 | 197 | 76 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Hamp| Gene Symbol: Hamp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004858837.3 | XP_004858894.1 | 1 - 2 | 208 | 76 | |
| GenBank mRNA | KM486717.1 | - | 1 - 2 | 197 | 76 | |