Assay Details
| Gene Symbol: Nrgn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004874462.3 | XP_004874519.1 | 2 - 3 | 283 | 62 | |
| XM_021251905.1 | XP_021107564.1 | 1 - 2 | 197 | 62 | ||
| XM_021251906.1 | XP_021107565.1 | 1 - 2 | 224 | 62 | ||
| GenBank mRNA | KM486742.1 | - | 1 - 2 | 103 | 62 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_004874462.3, XM_021251905.1, XM_021251906.1 transcripts.
Set Membership: |
|

Nrgn
Genomic Map