Assay Details
| Gene Symbol: Nrgn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004874462.3 | XP_004874519.1 | 3 - 4 | 509 | 94 | |
| XM_021251906.1 | XP_021107565.1 | 2 - 3 | 450 | 94 | ||
| GenBank mRNA | KM486742.1 | - | 2 - 3 | 329 | 94 | |

Nrgn
Genomic Map