Assay Details
| Gene Symbol: LOC101723866 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004875458.2 | XP_004875515.1 | 1 | 424 | 81 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_004875458.2 transcripts.
Set Membership: |
|

LOC101723866
Genomic Map